Anti-GFP Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: Rockland Immunochemicals
600-105-215
ROCK600-105-215EA 1310 EUR
ROCK600-105-215
Anti-GFP Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Anti-GFP is designed to detect GFP and its variants. This antibody can be used to detect GFP by ELISA (sandwich or capture) for the direct binding of antigen and recognizes wild type, recombinant and enhanced forms of GFP.
Recommended Dilutions: ELISA: 1:2000 - 1:8000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!
Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Full length
Host: Goat
Isotype: IgG
Reactivity: pan