Anti-GFP Goat Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals
600-106-215
ROCK600-106-215EA 1120 EUR
ROCK600-106-215
Anti-GFP Goat Polyclonal Antibody (Biotin)
Antikörper
GFP Antibody is suitable for immunoblotting, ELISA, immunochemistry, immunomicroscopy as well as other antibody based assays using streptavidin or avidin conjugates requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Full length
Host: Goat
Isotype: IgG
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR