Anti-RPE Goat Polyclonal Antibody (Biotin)
Lieferant: Rockland Immunochemicals
600-106-387
ROCK600-106-387EA 1010 EUR
ROCK600-106-387
Anti-RPE Goat Polyclonal Antibody (Biotin)
Antikörper
Suitable for immunoblotting (western or dot blot), ELISA as well as other immunochemical assays.  R-Phycoerythrin (240 kDa) is a labile molecule that may dissociate into components upon exposure to reducing or denaturing agents.
Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:2000 - 1:20000; Immunochemistry: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Tag
Antigen: RPE
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen