Anti-Biotin Goat Polyclonal Antibody (Texas Red®)

Lieferant: Rockland Immunochemicals
600-109-098
ROCK600-109-098EA 1120 EUR
ROCK600-109-098
Anti-Biotin Goat Polyclonal Antibody (Texas Red®)
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: Immunofluorescence: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR