Anti-Biotin Goat Polyclonal Antibody (DyLight® 488)

Lieferant: Rockland Immunochemicals
600-141-098
ROCK600-141-098EA 774 EUR
ROCK600-141-098
Anti-Biotin Goat Polyclonal Antibody (DyLight® 488)
Antikörper
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR