Anti-HSA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
600-401-033
ROCK600-401-033EA 2060 EUR
ROCK600-401-033
Anti-HSA Rabbit Polyclonal Antibody
Antikörper
Anti-Human Serum Albumin Antibody is suitable for the detection of human serum albumin in blood and tissue samples. Antibody is suitable for western blotting to detect a single band of 67 kDa, the expected apparent molecular weight, and for use in ELISA.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Primary
Antigen: HSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR