Anti-RFP Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-401-379
ROCK600-401-379EA 566 EUR
ROCK600-401-379
Anti-RFP Rabbit Polyclonal Antibody
Antikörper
Polyclonal anti-RFP is designed to detect RFP and its variants. This antibody can be used to detect RFP by ELISA (sandwich or capture) for the direct binding of antigen.
Recommended Dilutions: ELISA: 1:50000 - 1:100000; Immunofluorescence: 1:200-1:2000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:200 - 1:2000;
Type: Tag
Antigen: RFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen