Anti-6xHis Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-401-382
ROCK600-401-382EA 566 EUR
ROCK600-401-382
Anti-6xHis Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-6X HIS EPITOPE TAG Reacts with Pan
Type: Tag
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan