Anti-HA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
600-401-384
ROCK600-401-384EA 566 EUR
ROCK600-401-384
Anti-HA Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-HA EPITOPE TAG Reacts with Pan

Type: Tag
Antigen: HA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 114 - 122 aa
Host: Rabbit
Isotype: IgG
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR