Anti-RAD9 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
600-401-467
ROCK600-401-467EA 821 EUR
ROCK600-401-467
Anti-RAD9 Rabbit Polyclonal Antibody
Antikörper
This affinity purified antibody has been tested for use in ELISA against the immunizing peptide.  Reactivity in other immunoassays is unknown. Mature RAD9 is reported to have an apparent molecular weight of ~140kDa.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000;

Type: Primary
Antigen: RAD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae, Yeast
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR