Anti-NEU2 Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-401-850
ROCK600-401-850EA 821 EUR
ROCK600-401-850
Anti-NEU2 Rabbit Polyclonal Antibody
Antikörper
This antibody is suitable for western blotting,.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000;
Type: Primary
Antigen: NEU2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human