Anti-HSP90AA1 Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-401-981S
ROCK600-401-981SEA 226 EUR
ROCK600-401-981S
Anti-HSP90AA1 Rabbit Polyclonal Antibody
Antikörper
Anti-HSP 90 K294 has been tested for use in ELISA, immunohistochemistry and western blot.  This antibody reacts strongly with the acetylated form of the immunizing peptide and shows minimal reactivity with the non-acetylated form when tested by ELISA.
Recommended Dilutions: ELISA: 1:2500 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 5-10 µg/ml
Type: Primary
Antigen: HSP90AA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human