Anti-RRM2B Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
600-401-B67S
ROCK600-401-B67SEA 226 EUR
ROCK600-401-B67S
Anti-RRM2B Rabbit Polyclonal Antibody
Antikörper
RRM2B/p53R2 antibody is suitable for IP, IHC, IF, ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 41 kDa in size corresponding to RRM2B1 by western blotting.

Recommended Dilutions: ELISA: 1:3000; Western Blot: 1 µg/ml;

Type: Primary
Antigen: RRM2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR