Anti-BrdU Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-401-C29S
ROCK600-401-C29SEA 226 EUR
ROCK600-401-C29S
Anti-BrdU Rabbit Polyclonal Antibody
Antikörper
This affinity purified antibody has been tested as suitable for immunofluorescence microscopy. Antibody may be suitable for additional immunoassays including western blot, flow cytometry and immunochemistry.
Recommended Dilutions: ELISA: 1:2000-1:10000;
Type: Primary
Antigen: BrdU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen