Anti-GRIN1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
600-401-D85
ROCK600-401-D85EA 976 EUR
ROCK600-401-D85
Anti-GRIN1 Rabbit Polyclonal Antibody
Antikörper
Anti-NMDA R1, Splice Variant C1 (Rabbit) Antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 120 kDa in size corresponding to the NR1 subunit.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:1000-2000;
0.05 M Tris Chloride, 0.15 M Sodium Chloride, pH 7.4;

Type: Primary
Antigen: GRIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR