Anti-GRIN2C Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-401-D94
ROCK600-401-D94EA 976 EUR
ROCK600-401-D94
Anti-GRIN2C Rabbit Polyclonal Antibody
Antikörper
Anti-NMDA R2C Antibody is suitable for use in Western Blotting, Immunoprecipitation, IHC and IF. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 140 kDa in size corresponding to the NR2C subunit.
Recommended Dilutions: Immunofluorescence: 1:1000-2000; Western Blot: 1:1000; Immunochemistry: 1:1000-2000; Immunoprecipitation: 3µl per 200µg lysate;
Type: Primary
Antigen: GRIN2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat