Anti-PCLO Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-401-G30
ROCK600-401-G30EA 763 EUR
ROCK600-401-G30
Anti-PCLO Rabbit Polyclonal Antibody
Antikörper
Anti-Picollo Antibody is suitable for use in WB and IHC. Expect a band approximately ~550kDa corresponding to the molecular weight of Picollo. Multiple isoforms can be detected. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: PCLO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human