Anti-HIST2H3C Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
600-401-I55
ROCK600-401-I55EA 856 EUR
ROCK600-401-I55
Anti-HIST2H3C Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: ChIP: 2-5µg/million cells; Immunofluorescence: 1:50; Western Blot: 1:500; Immunochemistry: 1:50; Other Dilution: Dot Blot 1:1000

Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, C. elegans, Rat, Chicken, Xenopus, Drosophila, Plant
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR