Anti-HIST2H3C Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-401-I63
ROCK600-401-I63EA 856 EUR
ROCK600-401-I63
Anti-HIST2H3C Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: ChIP: 2-5µg/million cells; Immunofluorescence: 1:200; Western Blot: 1:500; Immunochemistry: 1:200; Other Dilution: Dot Blot user optimized
Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, C. elegans, Rat, Chicken, Xenopus, Drosophila, Plant