Anti-HIST2H3C Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-401-I95
ROCK600-401-I95EA 856 EUR
ROCK600-401-I95
Anti-HIST2H3C Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: ChIP: 2-5µg/million cells; Immunofluorescence: 1:100-1:1000; Western Blot: 1µg/ml; Immunochemistry: 1:100-1:1000; Other Dilution: user optimized
Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, C. elegans, Rat, Chicken, Xenopus, Drosophila, Plant