Anti-Biotin Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))

Lieferant: Rockland Immunochemicals
600-402-098
ROCK600-402-098EA 682 EUR
ROCK600-402-098
Anti-Biotin Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: Immunofluorescence: 1:500-1:2500
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR