Anti-GST Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))

Lieferant: Rockland Immunochemicals
600-402-200
ROCK600-402-200EA 774 EUR
ROCK600-402-200
Anti-GST Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Antikörper
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR