Anti-GFP Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))

Lieferant: Rockland Immunochemicals
600-402-215
ROCK600-402-215EA 774 EUR
ROCK600-402-215
Anti-GFP Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:20000-1:40000; Immunofluorescence: 1:500 - 1:2500; Flow Cytometry: 1:2000 suggested for starting optimization; Western Blot: 1:10000-1:25000
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR