Anti-RFP Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))

Lieferant: Rockland Immunochemicals
600-402-379
ROCK600-402-379EA 781 EUR
ROCK600-402-379
Anti-RFP Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Antikörper
Polyclonal anti-RFP is designed to detect RFP and its variants. This antibody can be used to detect RFP by ELISA (sandwich or capture) for the direct binding of antigen.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: 1:500 - 1:2500; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: RFP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR