Anti-GDF15 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Rockland Immunochemicals
600-403-B07
ROCK600-403-B07EA 774 EUR
ROCK600-403-B07
Anti-GDF15 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
This affinity purified antibody is suitable for ELISA and western blotting of human NAG-1 Protein.

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR