Anti-Y Rabbit Polyclonal Antibody (Biotin)
Lieferant: Rockland Immunochemicals
600-406-262
ROCK600-406-262EA 861 EUR
ROCK600-406-262
Anti-Y Rabbit Polyclonal Antibody (Biotin)
Antikörper
Anti-PhosphoTyrosine antibody is suitable for use in ELISA and western blotting assays.
Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:1000;
Type: Primary
Antigen: Y
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan