Anti-T Rabbit Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals
600-406-263
ROCK600-406-263EA 861 EUR
ROCK600-406-263
Anti-T Rabbit Polyclonal Antibody (Biotin)
Antikörper
This affinity-purified antibody is suitable for use in ELISA and western blotting assays.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:500;

Type: Primary
Antigen: T
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Phosphothreonine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR