Anti-HA Rabbit Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals
600-406-384
ROCK600-406-384EA 742 EUR
ROCK600-406-384
Anti-HA Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Tag
Antigen: HA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR