Anti-KLH Rabbit Polyclonal Antibody (Texas Red®)

Lieferant: Rockland Immunochemicals
600-409-466
ROCK600-409-466EA 774 EUR
ROCK600-409-466
Anti-KLH Rabbit Polyclonal Antibody (Texas Red®)
Antikörper
Recommended Dilutions: Immunofluorescence: 1:500 - 1:2500
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR