Anti-VSV-G Rabbit Polyclonal Antibody (DyLight® 488)

Lieferant: Rockland Immunochemicals
600-441-386
ROCK600-441-386EA 774 EUR
ROCK600-441-386
Anti-VSV-G Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Tag
Antigen: VSV-G
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR