Anti-6xHis Rabbit Polyclonal Antibody (DyLight® 800)
Lieferant: Rockland Immunochemicals
600-445-382
ROCK600-445-382EA 742 EUR
ROCK600-445-382
Anti-6xHis Rabbit Polyclonal Antibody (DyLight® 800)
Antikörper
Type: Tag
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan