Anti-FLAG™ conjugated Proteins Rabbit Polyclonal Antibody (DyLight® 800)

Lieferant: Rockland Immunochemicals
600-445-383
ROCK600-445-383EA 742 EUR
ROCK600-445-383
Anti-FLAG™ conjugated Proteins Rabbit Polyclonal Antibody (DyLight® 800)
Antikörper
Type: Primary
Antigen: FLAG™ conjugated Proteins
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR