Anti-GFP Chicken Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-901-215
ROCK600-901-215EA 603 EUR
ROCK600-901-215
Anti-GFP Chicken Polyclonal Antibody
Antikörper
Polyclonal anti-GFP is designed to detect GFP and its variants. This antibody can be used to detect GFP by ELISA (sandwich or capture) for the direct binding of antigen and recognizes wild type, recombinant and enhanced forms of GFP.
Recommended Dilutions: ELISA: 1:2000 - 1:12000; Western Blot: 1:200 - 1:1000; Immunochemistry: 1:200 - 1:1000;
Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Chicken
Isotype: IgG
Reactivity:
Bestellen