Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
602-4104
ROCK602-4104EA 253 EUR
ROCK602-4104
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype:
Reactivity: Cat