Anti-IgG (H+L) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Rockland Immunochemicals
602-4302
ROCK602-4302EA 290 EUR
ROCK602-4302
Anti-IgG (H+L) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Secondary Rabbit Anti-IgG (H&L) Reacts with Cat (Feline)
Recommended Dilutions: ELISA: 1:90,000; Western Blot: 1:5000 - 1:20000; Immunochemistry: 1:2000 - 1:7500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Cat