Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 649)

Lieferant: Rockland Immunochemicals
603-143-002-05
ROCK603-143-002-05EA 690 EUR
ROCK603-143-002-05
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 649)
Antikörper
Type: Secondary
Antigen: IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 649
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR