Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 680)
Lieferant: Rockland Immunochemicals
603-144-126
ROCK603-144-126EA 220 EUR
ROCK603-144-126
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 680)
Antikörper
The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000; Other Dilution: User Optimized
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 680
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Chicken