Anti-IgG (H+L) Donkey Polyclonal Antibody

Lieferant: Rockland Immunochemicals
603-701-C37
ROCK603-701-C37EA 253 EUR
ROCK603-701-C37
Anti-IgG (H+L) Donkey Polyclonal Antibody
Antikörper
Anti-Chicken IgY (H&L) is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays. Specific conditions for reactivity and signal detection should be optimized by the end user.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR