Anti-IgG F(c) Goat Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals
604-1603
ROCK604-1603EA 358 EUR
ROCK604-1603
Anti-IgG F(c) Goat Polyclonal Antibody (Biotin)
Antikörper
Anti-DOG IgG F(c) (GOAT) Antibody has been assayed against 1.0 µg of Dog IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR