Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 488)

Lieferant: Rockland Immunochemicals
605-441-013
ROCK605-441-013EA 220 EUR
ROCK605-441-013
Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR