Anti-IgG (H+L) Rabbit Polyclonal Antibody (ATTO 532)
Lieferant: Rockland Immunochemicals
605-453-013S
ROCK605-453-013SEA 237 EUR
ROCK605-453-013S
Anti-IgG (H+L) Rabbit Polyclonal Antibody (ATTO 532)
Antikörper
The emission spectra for this ATTO conjugate matches the principle output wavelengths of most common fluorescence instrumentation.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: ATTO 532
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Goat