Anti-IgG F(ab')2 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Rockland Immunochemicals
608-1204
ROCK608-1204EA 299 EUR
ROCK608-1204
Anti-IgG F(ab')2 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Secondary Goat Anti-IgG F(ab')2 Reacts with Horse (Equine)

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Horse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR