Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 488)

Lieferant: Rockland Immunochemicals
609-141-002-05
ROCK609-141-002-05EA 753 EUR
ROCK609-141-002-05
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Secondary
Antigen: IgG(Human)-HL
Clonality: Polyclonal
Clone:
Conjugation: DyLight488
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR