Anti-IgM µ Goat Polyclonal Antibody (DyLight® 549)
Lieferant: Rockland Immunochemicals
609-142-007
ROCK609-142-007EA 213 EUR
ROCK609-142-007
Anti-IgM µ Goat Polyclonal Antibody (DyLight® 549)
Antikörper
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope: µ-chain
Host: Goat
Isotype:
Reactivity: Human