Anti-IgM µ Goat Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals
609-1607
ROCK609-1607EA 308 EUR
ROCK609-1607
Anti-IgM µ Goat Polyclonal Antibody (Biotin)
Antikörper
Anti-HUMAN IgM (mu chain) (GOAT) Antibody has been assayed against 1.0 µg of Human IgM in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR