Anti-IgM Fc5µ Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
609-4131
ROCK609-4131EA 308 EUR
ROCK609-4131
Anti-IgM Fc5µ Rabbit Polyclonal Antibody
Antikörper
Human IgM Fc5µ antibody is suitable for immunoblotting (western or dot blot), ELISA, and immunochemistry requiring extremely low background levels, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:40000 - 1:160,000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:500 - 1:3000;

Type: Secondary
Antigen: IgM Fc5µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Fc5µ
Host: Rabbit
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR