Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 680)

Lieferant: Rockland Immunochemicals
609-444-002-05
ROCK609-444-002-05EA 753 EUR
ROCK609-444-002-05
Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 680)
Antikörper
Type: Secondary
Antigen: IgG(Human)-HL
Clonality: Polyclonal
Clone:
Conjugation: DyLight680
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR