Anti-IgM Fc5µ Rabbit Polyclonal Antibody (Texas Red®)

Lieferant: Rockland Immunochemicals
609-4931
ROCK609-4931EA 384 EUR
ROCK609-4931
Anti-IgM Fc5µ Rabbit Polyclonal Antibody (Texas Red®)
Antikörper
Secondary Rabbit Anti-IgM (Fc5u) Reacts with Human

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgM Fc5µ
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Fc5µ
Host: Rabbit
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR