Anti-IgG/IgM (H+L) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: Rockland Immunochemicals
610-100-115
ROCK610-100-115EA 319 EUR
ROCK610-100-115
Anti-IgG/IgM (H+L) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
A working dilution of 1:15,000 to 1:70,000 of the concentration is suggested for this product. Mouse IgM antibody is suitable for immunosassays including western blot and ELISA.

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR