Anti-IgG2b Goat Polyclonal Antibody (Texas Red®)

Lieferant: Rockland Immunochemicals
610-109-042
ROCK610-109-042EA 384 EUR
ROCK610-109-042
Anti-IgG2b Goat Polyclonal Antibody (Texas Red®)
Antikörper
Anti-Mouse IgG2b Texas Red® Conjugated antibodies are suitable for ELISA, immunochemistry, western blotting, immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting.

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Western Blot: 1:4000 - 1:20000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent);

Type: Secondary
Antigen: IgG2b γ
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR